Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KMT2A
Entrez Gene ID
4297
Gene Name
lysine methyltransferase 2A
Gene Aliases
ALL-1, CXXC7, HRX, HTRX1, MLL, MLL-AF9, MLL/GAS7, MLL1, MLL1A, TET1-MLL, TRX1, WDSTS
Chromosome Location
Chr.11: 118436490 - 118526832 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197104.1 NP_001184033.1 35 - 36 11669 75
NM_005933.3 NP_005924.2 35 - 36 11660 75
XM_006718839.3 XP_006718902.2 35 - 36 9144 75
XM_011542829.2 XP_011541131.1 36 - 37 11760 75
XM_011542830.2 XP_011541132.1 36 - 37 11757 75
XM_011542831.2 XP_011541133.1 36 - 37 11751 75
XM_011542833.2 XP_011541135.1 36 - 37 9243 75
GenBank mRNA AB209508.1 - 33 - 34 9204 75
AK295635.1 - 9 - 10 937 75
L04284.1 - 35 - 36 11637 75
L04731.1 - 35 - 36 11460 75

View Full Product Details