Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF19
Entrez Gene ID
124602
Gene Name
kinesin family member 19
Gene Aliases
KIF19A
Chromosome Location
Chr.17: 74326212 - 74355820 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153209.3 NP_694941.2 4 - 5 451 65
XM_005257029.1 XP_005257086.1 4 - 5 451 65
XM_005257030.1 XP_005257087.1 4 - 5 451 65
XM_017024150.1 XP_016879639.1 4 - 5 451 65
XM_017024151.1 XP_016879640.1 4 - 5 451 65
XM_017024152.1 XP_016879641.1 4 - 5 451 65
XM_017024153.1 XP_016879642.1 4 - 5 451 65
XM_017024154.1 XP_016879643.1 4 - 5 451 65
XM_017024155.1 XP_016879644.1 4 - 5 451 65
XM_017024156.1 XP_016879645.1 4 - 5 451 65
XM_017024157.1 XP_016879646.1 4 - 5 451 65
XM_017024158.1 XP_016879647.1 4 - 5 451 65
GenBank mRNA AK094619.1 - 4 - 5 406 65
BC026362.1 - 4 - 5 432 65
BC093631.1 - 4 - 5 337 65
BC104852.1 - 4 - 5 337 65
BC110989.1 - 4 - 5 415 65
BC143327.1 - 4 - 5 338 65
DA955730.1 - 4 - 5 451 65

View Full Product Details