The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153209.3 | NP_694941.2 | 6 - 7 | 720 | 62 |
XM_005257029.1 | XP_005257086.1 | 6 - 7 | 720 | 62 | |
XM_017024150.1 | XP_016879639.1 | 6 - 7 | 720 | 62 | |
XM_017024151.1 | XP_016879640.1 | 6 - 7 | 720 | 62 | |
XM_017024155.1 | XP_016879644.1 | 6 - 7 | 720 | 62 | |
XM_017024156.1 | XP_016879645.1 | 6 - 7 | 720 | 62 | |
GenBank mRNA | AK094619.1 | - | 6 - 7 | 675 | 62 |
BC093631.1 | - | 6 - 7 | 606 | 62 | |
BC104852.1 | - | 6 - 7 | 606 | 62 | |
BC110989.1 | - | 6 - 7 | 684 | 62 |