Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF19
Entrez Gene ID
124602
Gene Name
kinesin family member 19
Gene Aliases
KIF19A
Chromosome Location
Chr.17: 74326212 - 74355820 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153209.3 NP_694941.2 7 - 8 917 102
XM_005257029.1 XP_005257086.1 7 - 8 917 102
XM_005257030.1 XP_005257087.1 6 - 7 791 102
XM_017024150.1 XP_016879639.1 7 - 8 917 102
XM_017024151.1 XP_016879640.1 7 - 8 917 102
XM_017024152.1 XP_016879641.1 6 - 7 791 102
XM_017024153.1 XP_016879642.1 6 - 7 791 102
XM_017024154.1 XP_016879643.1 6 - 7 791 102
GenBank mRNA AK094619.1 - 7 - 8 872 102
BC026362.1 - 6 - 7 772 102
BC093631.1 - 7 - 8 803 102
BC104852.1 - 7 - 8 803 102
BC110989.1 - 7 - 8 881 102
BC143327.1 - 6 - 7 678 102

View Full Product Details