Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAPD7
Entrez Gene ID
11044
Gene Name
PAP associated domain containing 7
Gene Aliases
LAK-1, LAK1, POLK, POLS, TRF4, TRF4-1, TRF41, TUTASE5
Chromosome Location
Chr.5: 6713952 - 6757048 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171806.1 NP_001165277.1 6 - 7 1167 63
NM_006999.4 NP_008930.1 12 - 13 1565 63
XM_005248234.3 XP_005248291.1 12 - 13 2218 63
XM_011513945.1 XP_011512247.1 10 - 11 1341 63
XM_017008988.1 XP_016864477.1 12 - 13 2839 63
XM_017008989.1 XP_016864478.1 12 - 13 1477 63
XM_017008990.1 XP_016864479.1 7 - 8 2023 63
GenBank mRNA AB005754.3 - 11 - 12 1473 63
AK289857.1 - 11 - 12 1567 63
AK303516.1 - 6 - 7 1123 63
AK308405.1 - 6 - 7 1167 63
BC084567.1 - 12 - 13 1498 63
BC117137.1 - 11 - 12 1483 63
BC126106.1 - 11 - 12 1483 63
KC424495.1 - 12 - 13 2126 63

View Full Product Details