Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC39A8
Entrez Gene ID
64116
Gene Name
solute carrier family 39 member 8
Gene Aliases
BIGM103, CDG2N, LZT-Hs6, PP3105, ZIP8
Chromosome Location
Chr.4: 102251041 - 102345498 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135146.1 NP_001128618.1 8 - 9 1638 86
NM_001135148.1 NP_001128620.1 7 - 8 1075 86
NM_022154.5 NP_071437.3 7 - 8 1760 86
XM_005263177.1 XP_005263234.1 8 - 9 1547 86
XM_011532182.1 XP_011530484.1 6 - 7 1573 86
XM_017008540.1 XP_016864029.1 8 - 9 1555 86
XM_017008541.1 XP_016864030.1 10 - 11 1932 86
GenBank mRNA AB020970.1 - 2 - 3 226 86
AB040120.1 - 7 - 8 1708 86
AF193052.1 - 8 - 9 1589 86
AK027652.1 - 7 - 8 1075 86
BC001320.1 - 1 - 2 142 86
BC012125.1 - 8 - 9 1614 86

View Full Product Details