The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016323.3 | NP_057407.2 | 13 - 14 | 1887 | 120 |
GenBank mRNA | AB027289.1 | - | 13 - 14 | 1847 | 120 |
AK093155.1 | - | 3 - 4 | 1222 | 120 | |
AK301756.1 | - | 3 - 4 | 455 | 120 | |
AK302142.1 | - | 7 - 8 | 759 | 120 | |
AY337518.1 | - | 13 - 14 | 1887 | 120 | |
BC140716.1 | - | 13 - 14 | 1780 | 120 |