Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD4B
Entrez Gene ID
55095
Gene Name
sterile alpha motif domain containing 4B
Gene Aliases
SMGB, Smaug2
Chromosome Location
Chr.19: 39342459 - 39390748 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303614.1 NP_001290543.1 6 - 7 1427 69
NM_018028.3 NP_060498.2 9 - 10 2148 69
XM_011527057.2 XP_011525359.1 7 - 8 1438 69
XM_011527060.2 XP_011525362.1 7 - 8 2540 69
XM_011527061.2 XP_011525363.1 6 - 7 1352 69
XM_011527062.2 XP_011525364.1 6 - 7 1413 69
XM_011527063.2 XP_011525365.1 7 - 8 1437 69
XM_011527064.2 XP_011525366.1 7 - 8 1438 69
XM_017026918.1 XP_016882407.1 6 - 7 1372 69
XM_017026919.1 XP_016882408.1 8 - 9 1528 69
XM_017026920.1 XP_016882409.1 8 - 9 1550 69
XM_017026921.1 XP_016882410.1 6 - 7 1413 69
XM_017026922.1 XP_016882411.1 8 - 9 1530 69
XM_017026923.1 XP_016882412.1 8 - 9 7068 69
GenBank mRNA AK001073.1 - - 313 69
BC039652.1 - 2 - 3 125 69
BC054518.1 - 9 - 10 2139 69
BC065211.1 - 3 - 4 435 69
BC080641.1 - 6 - 7 1387 69
EF601121.1 - 9 - 10 2135 69

View Full Product Details