The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207011.1 | NP_001193940.1 | 2 - 3 | 173 | 85 |
NM_147164.2 | NP_671693.1 | 2 - 3 | 297 | 85 | |
XM_017014263.1 | XP_016869752.1 | 2 - 3 | 271 | 85 | |
XM_017014264.1 | XP_016869753.1 | 2 - 3 | 815 | 85 | |
GenBank mRNA | AL537375.3 | - | 2 - 3 | 172 | 85 |
M73238.1 | - | 2 - 3 | 266 | 85 |