The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291018.1 | NP_001277947.1 | 1 - 2 | 261 | 60 |
NM_002601.3 | NP_002592.1 | 1 - 2 | 261 | 60 | |
GenBank mRNA | AF022912.1 | - | 1 - 2 | 198 | 60 |
BC007831.2 | - | 1 - 2 | 197 | 60 | |
BT007278.1 | - | 1 - 2 | 48 | 60 | |
CR456980.1 | - | 1 - 2 | 48 | 60 |