The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005271972.4 | XP_005272029.1 | 7 - 8 | 563 | 76 |
XM_011543349.2 | XP_011541651.1 | 8 - 9 | 694 | 76 |