Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAIP
Entrez Gene ID
10293
Gene Name
TRAF interacting protein
Gene Aliases
RNF206, SCKL9, TRIP
Chromosome Location
Chr.3: 49828595 - 49856584 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005879.2 NP_005870.2 11 - 12 1147 108
XM_011533264.1 XP_011531566.1 11 - 12 1241 108
XM_017005526.1 XP_016861015.1 8 - 9 875 108
GenBank mRNA AB451320.1 - 11 - 12 108
AB451449.1 - 11 - 12 108
AK225873.1 - 8 - 9 826 108
AK292172.1 - 11 - 12 1135 108
BC000310.2 - 11 - 12 1128 108
BC019283.1 - 11 - 12 1128 108
U77845.1 - 11 - 12 1143 108

View Full Product Details