Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAIP
Entrez Gene ID
10293
Gene Name
TRAF interacting protein
Gene Aliases
RNF206, SCKL9, TRIP
Chromosome Location
Chr.3: 49828595 - 49856584 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005879.2 NP_005870.2 8 - 9 818 87
XM_011533264.1 XP_011531566.1 8 - 9 912 87
GenBank mRNA AB451320.1 - 8 - 9 712 87
AB451449.1 - 8 - 9 712 87
AK292172.1 - 8 - 9 806 87
BC000310.2 - 8 - 9 799 87
BC019283.1 - 8 - 9 799 87
U77845.1 - 8 - 9 814 87

View Full Product Details