The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308382.1 | NP_001295311.1 | 2 - 3 | 229 | 69 |
XM_006717777.1 | XP_006717840.1 | 1 - 2 | 950 | 69 | |
XM_011539664.1 | XP_011537966.1 | 1 - 2 | 950 | 69 | |
XM_011539665.2 | XP_011537967.1 | 2 - 3 | 538 | 69 | |
XM_011539666.1 | XP_011537968.1 | 2 - 3 | 216 | 69 | |
XM_017016093.1 | XP_016871582.1 | 2 - 3 | 475 | 69 | |
GenBank mRNA | BC003548.1 | - | 1 - 2 | 882 | 69 |
BI906881.1 | - | 2 - 3 | 189 | 69 |