Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35A5
Entrez Gene ID
55032
Gene Name
solute carrier family 35 member A5
Gene Aliases
-
Chromosome Location
Chr.3: 112561709 - 112584439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017945.3 NP_060415.1 4 - 5 647 146
XM_011512948.1 XP_011511250.1 4 - 5 497 146
GenBank mRNA AK000737.1 - 4 - 5 550 146
AK098839.1 - 4 - 5 560 146
AY358994.1 - 4 - 5 556 146
BC005207.1 - 4 - 5 609 146
BC010307.1 - 4 - 5 609 146
BC013046.1 - 4 - 5 551 146

View Full Product Details