The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021105.2 | NP_066928.1 | 5 - 6 | 747 | 152 |
XM_011512904.1 | XP_011511206.1 | 6 - 7 | 562 | 152 | |
XM_011512905.1 | XP_011511207.1 | 4 - 5 | 617 | 152 | |
XM_017006628.1 | XP_016862117.1 | 5 - 6 | 639 | 152 | |
XM_017006629.1 | XP_016862118.1 | 7 - 8 | 807 | 152 | |
GenBank mRNA | AB006746.1 | - | 5 - 6 | 615 | 152 |
AF098642.1 | - | 5 - 6 | 614 | 152 | |
AK313377.1 | - | 5 - 6 | 513 | 152 | |
BC021100.1 | - | 5 - 6 | 447 | 152 | |
BC032718.1 | - | 5 - 6 | 495 | 152 |