The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317996.1 | NP_001304925.1 | 3 - 4 | 243 | 80 |
NM_014337.3 | NP_055152.1 | 3 - 4 | 243 | 80 | |
NM_148175.2 | NP_680480.1 | 3 - 4 | 243 | 80 | |
NM_148176.2 | NP_680481.1 | 3 - 4 | 243 | 80 | |
XM_005261448.3 | XP_005261505.1 | 3 - 4 | 243 | 80 | |
XM_011530041.2 | XP_011528343.1 | 3 - 4 | 243 | 80 | |
XM_011530042.2 | XP_011528344.1 | 3 - 4 | 243 | 80 | |
XM_011530043.2 | XP_011528345.1 | 3 - 4 | 243 | 80 | |
XM_011530044.2 | XP_011528346.1 | 3 - 4 | 243 | 80 | |
XM_011530046.2 | XP_011528348.1 | 3 - 4 | 243 | 80 | |
XM_011530047.2 | XP_011528349.1 | 3 - 4 | 243 | 80 | |
XM_011530049.2 | XP_011528351.1 | 4 - 5 | 293 | 80 | |
XM_011530050.2 | XP_011528352.1 | 3 - 4 | 208 | 80 | |
XM_011530051.2 | XP_011528353.1 | 4 - 5 | 235 | 80 | |
GenBank mRNA | AK127259.1 | - | 4 - 5 | 266 | 80 |
AL702046.1 | - | 3 - 4 | 241 | 80 | |
BC000022.2 | - | 3 - 4 | 205 | 80 | |
BC028385.2 | - | 3 - 4 | 187 | 80 | |
CR456548.1 | - | 3 - 4 | 159 | 80 | |
DA863607.1 | - | 3 - 4 | 243 | 80 | |
U37219.1 | - | 3 - 4 | 219 | 80 |