Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC8A1
Entrez Gene ID
6546
Gene Name
solute carrier family 8 member A1
Gene Aliases
NCX1
Chromosome Location
Chr.2: 40112146 - 40512452 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112800.1 NP_001106271.1 7 - 8 2276 73
NM_001112801.1 NP_001106272.1 6 - 7 2252 73
NM_001112802.1 NP_001106273.1 6 - 7 2270 73
NM_001252624.1 NP_001239553.1 7 - 8 2267 73
NM_021097.2 NP_066920.1 8 - 9 2291 73
XM_005264514.3 XP_005264571.1 6 - 7 2365 73
XM_006712081.2 XP_006712144.1 9 - 10 2473 73
XM_006712082.3 XP_006712145.1 10 - 11 2575 73
XM_006712083.3 XP_006712146.1 9 - 10 2392 73
XM_006712084.3 XP_006712147.1 10 - 11 2673 73
XM_006712085.3 XP_006712148.1 9 - 10 3295 73
XM_011533050.1 XP_011531352.1 7 - 8 2267 73
XM_011533054.2 XP_011531356.1 9 - 10 2309 73
XM_011533055.2 XP_011531357.1 9 - 10 2436 73
XM_011533056.1 XP_011531358.1 9 - 10 2309 73
XM_011533057.2 XP_011531359.2 6 - 7 2352 73
XM_017004745.1 XP_016860234.1 9 - 10 2457 73
XM_017004746.1 XP_016860235.1 9 - 10 2310 73
XM_017004747.1 XP_016860236.1 7 - 8 2476 73
XM_017004748.1 XP_016860237.1 9 - 10 2378 73
XM_017004749.1 XP_016860238.1 10 - 11 2578 73
XM_017004750.1 XP_016860239.1 11 - 12 2812 73
XM_017004751.1 XP_016860240.1 11 - 12 2809 73
XM_017004752.1 XP_016860241.1 10 - 11 2613 73
XM_017004753.1 XP_016860242.1 9 - 10 2309 73
XM_017004754.1 XP_016860243.1 8 - 9 2413 73
XM_017004755.1 XP_016860244.1 7 - 8 2400 73
XM_017004756.1 XP_016860245.1 7 - 8 2480 73
XM_017004757.1 XP_016860246.1 7 - 8 2475 73
XM_017004758.1 XP_016860247.1 7 - 8 2454 73
XM_017004759.1 XP_016860248.1 7 - 8 2415 73
XM_017004760.1 XP_016860249.1 7 - 8 2389 73
XM_017004761.1 XP_016860250.1 7 - 8 2432 73
XM_017004762.1 XP_016860251.1 6 - 7 2386 73
XM_017004763.1 XP_016860252.1 6 - 7 2368 73
GenBank mRNA AB209075.1 - 7 - 8 581 73
AF108388.1 - 6 - 7 2228 73
AF108389.1 - 5 - 6 2159 73
AF128524.1 - 6 - 7 2250 73
AK291696.1 - 6 - 7 2270 73
BC098285.1 - 8 - 9 501 73
BC098308.1 - 7 - 8 485 73
BX648299.1 - 6 - 7 2350 73
M91368.1 - - 2347 73
M96368.1 - 8 - 9 2308 73

View Full Product Details