Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAH14
Entrez Gene ID
127602
Gene Name
dynein axonemal heavy chain 14
Gene Aliases
C1orf67, Dnahc14, HL-18, HL18
Chromosome Location
Chr.1: 224928422 - 225399294 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144989.2 NP_659426.2 6 - 7 860 131
GenBank mRNA AI076810.1 - 1 - 2 131
BC017415.2 - 5 - 6 686 131

View Full Product Details