The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000231.2 | NP_000222.1 | 4 - 5 | 541 | 119 |
XM_005266505.2 | XP_005266562.1 | 5 - 6 | 634 | 119 | |
XM_006719861.3 | XP_006719924.1 | 4 - 5 | 725 | 119 | |
GenBank mRNA | AK316579.1 | - | 5 - 6 | 609 | 119 |
BC074777.2 | - | 5 - 6 | 429 | 119 | |
BC074778.2 | - | 5 - 6 | 428 | 119 | |
BC109321.2 | - | 4 - 5 | 501 | 119 | |
DA897096.1 | - | 4 - 5 | 457 | 119 | |
U34976.1 | - | 5 - 6 | 510 | 119 |