Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PKP4
Entrez Gene ID
8502
Gene Name
plakophilin 4
Gene Aliases
p0071
Chromosome Location
Chr.2: 158456880 - 158681429 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005476.2 NP_001005476.1 14 - 15 2746 106
NM_001304969.1 NP_001291898.1 14 - 15 2743 106
NM_001304970.1 NP_001291899.1 14 - 15 2666 106
NM_001304971.1 NP_001291900.1 14 - 15 2746 106
NM_003628.4 NP_003619.2 14 - 15 2746 106
XM_011512016.1 XP_011510318.1 14 - 15 3183 106
XM_011512017.1 XP_011510319.1 14 - 15 2487 106
XM_011512018.1 XP_011510320.1 14 - 15 3177 106
XM_011512019.1 XP_011510321.1 14 - 15 2484 106
XM_011512020.2 XP_011510322.1 14 - 15 2530 106
XM_011512021.2 XP_011510323.1 14 - 15 2530 106
XM_011512022.1 XP_011510324.1 13 - 14 2698 106
XM_011512025.1 XP_011510327.1 9 - 10 1618 106
XM_017005122.1 XP_016860611.1 14 - 15 3180 106
XM_017005123.1 XP_016860612.1 14 - 15 3715 106
XM_017005124.1 XP_016860613.1 14 - 15 3174 106
XM_017005125.1 XP_016860614.1 14 - 15 2527 106
XM_017005126.1 XP_016860615.1 14 - 15 2487 106
XM_017005127.1 XP_016860616.1 14 - 15 3183 106
XM_017005128.1 XP_016860617.1 14 - 15 2527 106
XM_017005129.1 XP_016860618.1 13 - 14 2692 106
XM_017005130.1 XP_016860619.1 13 - 14 2695 106
XM_017005131.1 XP_016860620.1 9 - 10 1615 106
XM_017005132.1 XP_016860621.1 12 - 13 2023 106
XM_017005133.1 XP_016860622.1 13 - 14 2150 106
XM_017005134.1 XP_016860623.1 12 - 13 2517 106
XM_017005135.1 XP_016860624.1 11 - 12 1720 106
GenBank mRNA AK124237.1 - 1 - 2 846 106
AK128720.1 - 12 - 13 2573 106
BC050308.1 - 14 - 15 2528 106
X81889.1 - 14 - 15 2543 106

View Full Product Details