Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKP4
Entrez Gene ID
8502
Gene Name
plakophilin 4
Gene Aliases
p0071
Chromosome Location
Chr.2: 158456880 - 158681429 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005476.2 NP_001005476.1 6 - 7 944 77
NM_001304969.1 NP_001291898.1 6 - 7 944 77
NM_001304971.1 NP_001291900.1 6 - 7 944 77
NM_003628.4 NP_003619.2 6 - 7 944 77
XM_011512016.1 XP_011510318.1 6 - 7 1381 77
XM_011512017.1 XP_011510319.1 6 - 7 685 77
XM_011512018.1 XP_011510320.1 6 - 7 1375 77
XM_011512019.1 XP_011510321.1 6 - 7 685 77
XM_011512020.2 XP_011510322.1 6 - 7 728 77
XM_011512021.2 XP_011510323.1 6 - 7 728 77
XM_011512022.1 XP_011510324.1 5 - 6 896 77
XM_011512026.2 XP_011510328.1 6 - 7 728 77
XM_017005122.1 XP_016860611.1 6 - 7 1381 77
XM_017005123.1 XP_016860612.1 6 - 7 1916 77
XM_017005124.1 XP_016860613.1 6 - 7 1375 77
XM_017005125.1 XP_016860614.1 6 - 7 728 77
XM_017005126.1 XP_016860615.1 6 - 7 685 77
XM_017005127.1 XP_016860616.1 6 - 7 1381 77
XM_017005128.1 XP_016860617.1 6 - 7 728 77
XM_017005129.1 XP_016860618.1 5 - 6 890 77
XM_017005130.1 XP_016860619.1 5 - 6 896 77
GenBank mRNA AK055823.1 - - 1030 77
AK128720.1 - 4 - 5 771 77
BC050308.1 - 6 - 7 726 77
U60472.1 - 1 - 2 156 77
X81889.1 - 6 - 7 741 77

View Full Product Details