Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASGEF1C
Entrez Gene ID
255426
Gene Name
RasGEF domain family member 1C
Gene Aliases
-
Chromosome Location
Chr.5: 180100795 - 180209130 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175062.3 NP_778232.2 10 - 11 1191 132
GenBank mRNA AF449764.1 - 9 - 10 1379 132
AK093160.1 - 7 - 8 1043 132
BC036802.1 - 11 - 12 1209 132

View Full Product Details