The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003704.2 | NP_001003704.1 | 1 - 2 | 327 | 65 |
NM_016498.4 | NP_057582.2 | 1 - 2 | 327 | 65 | |
GenBank mRNA | AF060924.1 | - | 1 - 2 | 240 | 65 |
AK026104.1 | - | 1 - 2 | 195 | 65 | |
BC001608.1 | - | 1 - 2 | 153 | 65 | |
BC009300.2 | - | 1 - 2 | 185 | 65 | |
BC014446.1 | - | 1 - 2 | 153 | 65 | |
BC030989.2 | - | 1 - 2 | 214 | 65 | |
BC038831.2 | - | 1 - 2 | 218 | 65 | |
BC046132.1 | - | 1 - 2 | 117 | 65 | |
BQ187590.1 | - | 1 - 2 | 109 | 65 | |
CB107806.1 | - | 1 - 2 | 327 | 65 | |
CB142469.1 | - | 1 - 2 | 218 | 65 | |
CR456345.1 | - | 1 - 2 | 122 | 65 |