The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039582.3 | NP_001034671.3 | 9 - 10 | 1242 | 66 |
NM_001135740.1 | NP_001129212.1 | 9 - 10 | 876 | 66 | |
XM_005274651.2 | XP_005274708.1 | 9 - 10 | 859 | 66 | |
XM_006724809.3 | XP_006724872.1 | 9 - 10 | 1068 | 66 | |
XM_011531107.2 | XP_011529409.2 | 9 - 10 | 947 | 66 | |
XM_011531108.2 | XP_011529410.1 | 9 - 10 | 912 | 66 | |
XM_011531110.2 | XP_011529412.1 | 9 - 10 | 964 | 66 | |
XM_011531111.2 | XP_011529413.1 | 9 - 10 | 882 | 66 | |
XM_011531112.2 | XP_011529414.1 | 9 - 10 | 855 | 66 | |
XM_017029277.1 | XP_016884766.1 | 9 - 10 | 1167 | 66 | |
XM_017029278.1 | XP_016884767.1 | 10 - 11 | 1071 | 66 | |
XM_017029279.1 | XP_016884768.1 | 9 - 10 | 828 | 66 | |
GenBank mRNA | AK296165.1 | - | 8 - 9 | 1285 | 66 |
AK296203.1 | - | 9 - 10 | 1242 | 66 | |
AK303746.1 | - | 9 - 10 | 876 | 66 | |
BC064422.1 | - | 8 - 9 | 934 | 66 |