The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_120603.1 | 4 - 5 | 432 | 115 | |
GenBank mRNA | AJ251022.1 | - | 4 - 5 | 432 | 115 |