The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272003.1 | NP_001258932.1 | 22 - 23 | 2668 | 69 |
NM_020822.2 | NP_065873.2 | 23 - 24 | 2803 | 69 | |
XM_011518877.2 | XP_011517179.1 | 23 - 24 | 6773 | 69 | |
XM_011518878.2 | XP_011517180.1 | 23 - 24 | 6782 | 69 | |
XM_011518879.2 | XP_011517181.1 | 23 - 24 | 6773 | 69 | |
XM_011518880.1 | XP_011517182.1 | 22 - 23 | 2630 | 69 | |
XM_011518881.2 | XP_011517183.1 | 20 - 21 | 3690 | 69 | |
XM_017014931.1 | XP_016870420.1 | 21 - 22 | 2678 | 69 | |
XM_017014932.1 | XP_016870421.1 | 22 - 23 | 2570 | 69 | |
XM_017014933.1 | XP_016870422.1 | 20 - 21 | 2397 | 69 | |
GenBank mRNA | AB037843.1 | - | 23 - 24 | 2796 | 69 |
AK127272.1 | - | 22 - 23 | 2668 | 69 | |
BC136618.1 | - | 23 - 24 | 2775 | 69 | |
BC171770.1 | - | 23 - 24 | 2775 | 69 |