The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272003.1 | NP_001258932.1 | 7 - 8 | 603 | 97 |
NM_020822.2 | NP_065873.2 | 8 - 9 | 747 | 97 | |
XM_011518877.2 | XP_011517179.1 | 8 - 9 | 4717 | 97 | |
XM_011518878.2 | XP_011517180.1 | 8 - 9 | 4717 | 97 | |
XM_011518879.2 | XP_011517181.1 | 8 - 9 | 4717 | 97 | |
XM_011518880.1 | XP_011517182.1 | 7 - 8 | 574 | 97 | |
XM_011518881.2 | XP_011517183.1 | 5 - 6 | 1625 | 97 | |
XM_017014931.1 | XP_016870420.1 | 6 - 7 | 613 | 97 | |
XM_017014932.1 | XP_016870421.1 | 7 - 8 | 505 | 97 | |
XM_017014933.1 | XP_016870422.1 | 5 - 6 | 332 | 97 | |
GenBank mRNA | AB037843.1 | - | 8 - 9 | 740 | 97 |
AK127272.1 | - | 7 - 8 | 603 | 97 | |
BC136618.1 | - | 8 - 9 | 719 | 97 | |
BC171770.1 | - | 8 - 9 | 719 | 97 |