The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204744.1 | NP_001191673.1 | 6 - 7 | 779 | 126 |
NM_001204745.1 | NP_001191674.1 | 6 - 7 | 779 | 126 | |
NM_004062.3 | NP_004053.1 | 6 - 7 | 779 | 126 | |
XM_005255770.2 | XP_005255827.1 | 6 - 7 | 670 | 126 | |
XM_011522807.1 | XP_011521109.1 | 6 - 7 | 649 | 126 | |
GenBank mRNA | AF016272.1 | - | 6 - 7 | 622 | 126 |
AK225552.1 | - | 6 - 7 | 647 | 126 | |
AK298255.1 | - | 6 - 7 | 644 | 126 | |
AK298334.1 | - | 6 - 7 | 539 | 126 | |
AK313101.1 | - | 6 - 7 | 649 | 126 | |
AY358911.1 | - | 6 - 7 | 712 | 126 | |
BC027912.2 | - | 5 - 6 | 598 | 126 |