Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IRX1
Entrez Gene ID
79192
Gene Name
iroquois homeobox 1
Gene Aliases
IRX-5, IRXA1
Chromosome Location
Chr.5: 3595855 - 3601403 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024337.3 NP_077313.3 2 - 3 1370 110
GenBank mRNA AY335938.1 - 2 - 3 1370 110
BC029160.1 - 2 - 3 1072 110

View Full Product Details