The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085454.1 | NP_001078923.1 | 13 - 14 | 1468 | 111 |
NM_014030.3 | NP_054749.2 | 12 - 13 | 1441 | 111 | |
XM_011524684.1 | XP_011522986.1 | 13 - 14 | 1309 | 111 | |
XM_011524685.1 | XP_011522987.1 | 12 - 13 | 1282 | 111 | |
GenBank mRNA | AB209537.1 | - | 13 - 14 | 1267 | 111 |
AF124490.1 | - | 12 - 13 | 1241 | 111 | |
AK024826.1 | - | 7 - 8 | 596 | 111 | |
AK294785.1 | - | 14 - 15 | 1346 | 111 | |
AK297443.1 | - | 10 - 11 | 910 | 111 | |
AK299615.1 | - | 4 - 5 | 338 | 111 | |
AK299932.1 | - | 13 - 14 | 1312 | 111 | |
BC067358.1 | - | 12 - 13 | 1441 | 111 | |
BX537553.1 | - | 5 - 6 | 1017 | 111 | |
BX537554.1 | - | 5 - 6 | 1017 | 111 | |
BX648097.1 | - | 5 - 6 | 1020 | 111 |