The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085454.1 | NP_001078923.1 | 3 - 4 | 516 | 67 |
NM_014030.3 | NP_054749.2 | 3 - 4 | 516 | 67 | |
XM_011524684.1 | XP_011522986.1 | 3 - 4 | 357 | 67 | |
XM_011524685.1 | XP_011522987.1 | 3 - 4 | 357 | 67 | |
GenBank mRNA | AB209537.1 | - | 3 - 4 | 315 | 67 |
AF124490.1 | - | 3 - 4 | 316 | 67 | |
AK294785.1 | - | 4 - 5 | 394 | 67 | |
AK299932.1 | - | 3 - 4 | 360 | 67 | |
BC048196.1 | - | 3 - 4 | 543 | 67 | |
BC067358.1 | - | 3 - 4 | 516 | 67 | |
BQ424386.1 | - | 2 - 3 | 155 | 67 | |
DR001908.1 | - | 3 - 4 | 497 | 67 |