The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005258048.4 | XP_005258105.2 | 1 - 2 | 216 | 88 |
XM_011525385.2 | XP_011523687.1 | 1 - 2 | 216 | 88 | |
XM_011525386.2 | XP_011523688.1 | 1 - 2 | 216 | 88 | |
XM_011525387.2 | XP_011523689.1 | 1 - 2 | 216 | 88 | |
XM_011525388.2 | XP_011523690.1 | 1 - 2 | 216 | 88 | |
XM_011525389.2 | XP_011523691.1 | 1 - 2 | 216 | 88 |