Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RAPGEF1
- Entrez Gene ID
-
2889
- Gene Name
- Rap guanine nucleotide exchange factor 1
- Gene Aliases
-
C3G, GRF2
- Chromosome Location
-
Chr.9: 131576770 - 131740068 on Build GRCh38
View Full Product Details