Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AXIN2
Entrez Gene ID
8313
Gene Name
axin 2
Gene Aliases
AXIL, ODCRCS
Chromosome Location
Chr.17: 65528563 - 65561622 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004655.3 NP_004646.3 10 - 11 2689 80
XM_011525319.2 XP_011523621.1 10 - 11 2900 80
XM_011525320.1 XP_011523622.1 10 - 11 2889 80
XM_011525321.2 XP_011523623.1 10 - 11 2837 80
XM_017025192.1 XP_016880681.1 10 - 11 2556 80
XM_017025193.1 XP_016880682.1 9 - 10 2694 80
XM_017025194.1 XP_016880683.1 9 - 10 2473 80
GenBank mRNA AB052751.1 - 3 - 4 314 80
AF078165.1 - 9 - 10 2489 80
AF205888.1 - 8 - 9 2311 80
AK025718.1 - 9 - 10 1711 80
AK315303.1 - 10 - 11 2605 80
BC101533.1 - 9 - 10 2469 80
BC143244.1 - 8 - 9 2274 80
CR933657.1 - 10 - 11 2714 80
CX759580.1 - 1 - 2 69 80

View Full Product Details