The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018225.2 | NP_060695.2 | 8 - 9 | 1098 | 108 |
XM_005251503.4 | XP_005251560.1 | 7 - 8 | 934 | 108 | |
GenBank mRNA | AK001667.1 | - | 8 - 9 | 1028 | 108 |
AK225559.1 | - | 8 - 9 | 1053 | 108 | |
AK225587.1 | - | 8 - 9 | 1047 | 108 | |
AK304767.1 | - | 6 - 7 | 685 | 108 | |
BC002876.2 | - | 8 - 9 | 1044 | 108 | |
BC110654.1 | - | 8 - 9 | 1138 | 108 | |
EF011613.1 | - | 8 - 9 | 1044 | 108 |