Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MBNL2
Entrez Gene ID
10150
Gene Name
muscleblind like splicing regulator 2
Gene Aliases
MBLL, MBLL39, PRO2032
Chromosome Location
Chr.13: 97141780 - 97394120 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005254018.2 XP_005254075.1 5 - 6 1931 125
XM_005254019.2 XP_005254076.1 5 - 6 1931 125
XM_005254021.2 XP_005254078.1 5 - 6 1931 125
XM_006719912.2 XP_006719975.1 5 - 6 1931 125
XM_011521045.2 XP_011519347.1 5 - 6 1538 125
XM_017020308.1 XP_016875797.1 5 - 6 1538 125
GenBank mRNA AF061261.1 - 6 - 7 1551 88

View Full Product Details