The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142556.1 | NP_001136028.1 | 3 - 4 | 407 | 117 |
GenBank mRNA | AK302975.1 | - | 3 - 4 | 286 | 117 |
AK313603.1 | - | 3 - 4 | 325 | 117 | |
AY623426.1 | - | 3 - 4 | 230 | 117 | |
BC030505.2 | - | 3 - 4 | 314 | 117 | |
BC108904.1 | - | 3 - 4 | 259 | 117 | |
U29343.1 | - | 3 - 4 | 285 | 117 |