The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146040.1 | NP_001139512.1 | 8 - 9 | 1348 | 97 |
GenBank mRNA | BC114947.1 | - | 8 - 9 | 1348 | 97 |