The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019556.2 | NP_062456.1 | 4 - 5 | 705 | 150 |
XM_011531366.2 | XP_011529668.1 | 4 - 5 | 659 | 150 | |
XM_017029653.1 | XP_016885142.1 | 5 - 6 | 898 | 150 | |
GenBank mRNA | AK307196.1 | - | 3 - 4 | 532 | 150 |
AK316570.1 | - | 4 - 5 | 580 | 150 | |
AL137163.1 | - | 4 - 5 | 628 | 150 | |
BC005700.1 | - | 4 - 5 | 608 | 150 |