The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033554.2 | NP_001028726.1 | 1 - 2 | 241 | 57 |
NM_001033555.2 | NP_001028727.1 | 1 - 2 | 241 | 57 | |
NM_001243438.1 | NP_001230367.1 | 1 - 2 | 241 | 57 | |
XM_017025321.1 | XP_016880810.1 | 1 - 2 | 241 | 57 | |
GenBank mRNA | AK304186.1 | - | 1 - 2 | 163 | 57 |
AY816326.1 | - | 1 - 2 | 146 | 57 | |
AY816327.1 | - | 1 - 2 | 146 | 57 | |
BC021123.1 | - | 1 - 2 | 139 | 57 | |
BC050058.1 | - | 1 - 2 | 76 | 57 | |
DC336225.1 | - | 1 - 2 | 246 | 57 |