Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EVC
Entrez Gene ID
2121
Gene Name
EvC ciliary complex subunit 1
Gene Aliases
DWF-1, EVC1, EVCL
Chromosome Location
Chr.4: 5711197 - 5829043 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306090.1 NP_001293019.1 14 - 15 2286 89
NM_153717.2 NP_714928.1 14 - 15 2286 89
XM_006713865.3 XP_006713928.1 14 - 15 2283 89
XM_006713866.3 XP_006713929.1 14 - 15 2283 89
GenBank mRNA AF216184.1 - 14 - 15 2286 89
AF216185.1 - 14 - 15 2286 89
AF239742.1 - 14 - 15 2102 89

View Full Product Details