The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306090.1 | NP_001293019.1 | 16 - 17 | 2637 | 82 |
NM_153717.2 | NP_714928.1 | 16 - 17 | 2637 | 82 | |
XM_006713865.3 | XP_006713928.1 | 16 - 17 | 2634 | 82 | |
XM_006713866.3 | XP_006713929.1 | 16 - 17 | 2634 | 82 | |
GenBank mRNA | AF216184.1 | - | 16 - 17 | 2637 | 82 |
AF216185.1 | - | 16 - 17 | 2637 | 82 | |
AF239742.1 | - | 16 - 17 | 2453 | 82 |