The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306090.1 | NP_001293019.1 | 5 - 6 | 879 | 95 |
NM_001306092.1 | NP_001293021.1 | 5 - 6 | 879 | 95 | |
NM_153717.2 | NP_714928.1 | 5 - 6 | 879 | 95 | |
XM_006713865.3 | XP_006713928.1 | 5 - 6 | 876 | 95 | |
XM_006713866.3 | XP_006713929.1 | 5 - 6 | 876 | 95 | |
XM_011513419.2 | XP_011511721.1 | 5 - 6 | 873 | 95 | |
XM_017007883.1 | XP_016863372.1 | 5 - 6 | 873 | 95 | |
GenBank mRNA | AF216184.1 | - | 5 - 6 | 879 | 95 |
AF216185.1 | - | 5 - 6 | 879 | 95 | |
AF239742.1 | - | 5 - 6 | 695 | 95 | |
BC085608.1 | - | 5 - 6 | 861 | 95 |