Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN17
Entrez Gene ID
26262
Gene Name
tetraspanin 17
Gene Aliases
FBX23, FBXO23, TM4SF17
Chromosome Location
Chr.5: 176647387 - 176659058 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY037146.1 - 2 - 3 406 70

View Full Product Details