The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005236.2 | NP_005227.1 | 2 - 3 | 397 | 118 |
XM_017023043.1 | XP_016878532.1 | 2 - 3 | 363 | 118 | |
GenBank mRNA | AK289726.1 | - | 2 - 3 | 396 | 118 |
AK308341.1 | - | 2 - 3 | 363 | 118 | |
BC020741.1 | - | 2 - 3 | 368 | 118 | |
BC142631.1 | - | 2 - 3 | 397 | 118 | |
U64315.1 | - | 2 - 3 | 370 | 118 |