The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278506.1 | NP_001265435.1 | 5 - 6 | 499 | 78 |
NM_001323594.1 | NP_001310523.1 | 5 - 6 | 393 | 78 | |
NM_001323595.1 | NP_001310524.1 | 6 - 7 | 458 | 78 | |
NM_001323596.1 | NP_001310525.1 | 6 - 7 | 483 | 78 | |
NM_001323597.1 | NP_001310526.1 | 5 - 6 | 449 | 78 | |
NM_002484.3 | NP_002475.2 | 6 - 7 | 532 | 78 | |
XM_017023252.1 | XP_016878741.1 | 5 - 6 | 1085 | 78 | |
GenBank mRNA | AK223204.1 | - | 6 - 7 | 474 | 78 |
AK310089.1 | - | 6 - 7 | 477 | 78 | |
BC100290.1 | - | 5 - 6 | 442 | 78 | |
BC109322.1 | - | 6 - 7 | 458 | 78 | |
BC109323.1 | - | 6 - 7 | 458 | 78 | |
BP216107.1 | - | 6 - 7 | 477 | 78 | |
U01833.1 | - | 6 - 7 | 458 | 78 |