The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077350.2 | NP_001070818.1 | 4 - 5 | 441 | 107 |
NM_001243247.1 | NP_001230176.1 | 5 - 6 | 578 | 107 | |
GenBank mRNA | AK026990.1 | - | 1 - 2 | 121 | 107 |
AK294228.1 | - | 5 - 6 | 574 | 107 | |
DA258664.1 | - | 4 - 5 | 415 | 107 | |
DA658076.1 | - | 4 - 5 | 416 | 107 | |
X90857.1 | - | 4 - 5 | 602 | 107 |