Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR62
Entrez Gene ID
284403
Gene Name
WD repeat domain 62
Gene Aliases
C19orf14, MCPH2
Chromosome Location
Chr.19: 36054881 - 36111145 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083961.1 NP_001077430.1 17 - 18 2234 64
NM_173636.4 NP_775907.4 17 - 18 2234 64
XM_005258809.2 XP_005258866.1 17 - 18 2234 64
XM_011526837.1 XP_011525139.1 17 - 18 2219 64
XM_011526838.1 XP_011525140.1 17 - 18 2234 64
XM_011526839.1 XP_011525141.1 15 - 16 1883 64
XM_011526840.2 XP_011525142.1 10 - 11 1229 64
XM_011526841.2 XP_011525143.1 8 - 9 850 64
XM_011526842.1 XP_011525144.1 7 - 8 682 64
XM_017026665.1 XP_016882154.1 17 - 18 2234 64
GenBank mRNA BC058939.1 - 17 - 18 2178 64
BQ880028.1 - 4 - 5 403 64
BX647726.1 - 18 - 19 2295 64

View Full Product Details