Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMYD2
Entrez Gene ID
56950
Gene Name
SET and MYND domain containing 2
Gene Aliases
HSKM-B, KMT3C, ZMYND14
Chromosome Location
Chr.1: 214281101 - 214337134 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020197.2 NP_064582.2 4 - 5 435 109
GenBank mRNA AF226053.1 - 4 - 5 424 109
AK313868.1 - 4 - 5 405 109
BC028104.1 - 3 - 4 356 109
BC068511.1 - 3 - 4 501 109
BC098133.1 - 4 - 5 424 109
BC098276.1 - 4 - 5 424 109
BC098305.1 - 4 - 5 424 109
BC098335.1 - 4 - 5 424 109

View Full Product Details